missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrin beta-chain Antibody (59D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43554
This item is not returnable.
View return policy
Description
Fibrin beta-chain Monoclonal antibody specifically detects Fibrin beta-chain in Human samples. It is validated for ELISA
Specifications
| Fibrin beta-chain | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p | |
| The original antibody was generated by using a synthetic heptapeptide from the amino terminus of the beta chain in human fibrin as an immunogen. (Uniprot# P02675) | |
| 0.2 mg | |
| Cell Cycle and Replication | |
| 2244 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| 59D8 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction