missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FH Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82508-1-RR-20UL
This item is not returnable.
View return policy
Description
The FH antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FH. This antibody recognizes human, rat, mouse antigen. The FH antibody has been validated for the following applications: WB, IP, ELISA analysis.Specifications
| FH | |
| Recombinant Monoclonal | |
| 500 μg/mL | |
| HLRCC; LRCC; MCL; MCUL1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 45 kDa | |
| Liquid |
| Western Blot, Immunoprecipitation, ELISA | |
| 3K16 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 2271 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction