missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody (1E3F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | FGFR3 |
|---|---|
| Clone | 1E3F9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266580
|
Novus Biologicals
NBP2-52469 |
0.1 mg |
552.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698846
|
Novus Biologicals
NBP2-52469-0.025mg |
0.025 mg |
203.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
| FGFR3 | |
| 1 mg/mL | |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| 2261 | |
| Purified recombinant fragment of human FGFR3 (AA: 529-694) expressed in E. Coli. | |
| Primary | |
| 87.7 kDa |
| 1E3F9 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| FGFR3 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title