Learn More
FGF18 Antibody (CL7628) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88869PE
Product Code. 30308554
  • 737.00€ / 0.10mL
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF18
Monoclonal
PE
FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
This antibody was developed using a synthetic peptide derived from O76093, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication
8817
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL7628
PBS
Mouse
Protein A purified