missing translation for 'onlineSavingsMsg'
Learn More

FGF-22 Antibody (MM0286-4F28), CoraFluor™ 1, Novus Biologicals™

Product Code. 30156125
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30156125 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30156125 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP212300CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-22 Monoclonal antibody specifically detects FGF-22 in Human samples. It is validated for Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-22
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0286-4F28
Conjugate CoraFluor 1
Formulation PBS
Gene Alias FGF-22, fibroblast growth factor 22
Host Species Mouse
Immunogen Human recombinant FGF-22
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 27006
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.