missing translation for 'onlineSavingsMsg'
Learn More

FGF-22 Antibody (MM0286-4F28), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18141450 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18141450 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18141450 Supplier Novus Biologicals Supplier No. NBP212300AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-22 Monoclonal specifically detects FGF-22 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-22
Applications Western Blot
Classification Monoclonal
Clone MM0286-4F28
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FGF-22, fibroblast growth factor 22
Gene Symbols FGF22
Host Species Mouse
Immunogen Human recombinant FGF-22
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27006
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.