missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BDKRB2 Monoclonal Antibody (471916)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561479
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Bradykinin B2 receptor (BDKRB2) expression has been reported in blood, brain, eye, kidney, liver, lung, prostate, skin, small intestine, synovium, uterus, and vessel. ESTs have been isolated from colon, connective tissue, heart, heart/melanocyte/uterus, and kidney libraries. It is associated with G-proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications
| BDKRB2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P30411 | |
| BDKRB2 | |
| NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 471916 | |
| Unconjugated | |
| BDKRB2 | |
| B(2); B2; B2 bradykinin receptor; B2BKR; B2BRA; B2R; BDKRB2; BK2; BK-2; BK-2 receptor; BK2R; BK-2R; BK-2Rs; BKR2; bradykinin receptor B2; bradykinin receptor, beta 2; BRB2; DKFZp686O088; kinin B2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 624 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction