Learn More
Thermo Scientific™ Ferrules for Thermo Scientific™ and Agilent™ Instruments
Product Code. 10394473 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
100% Graphite
15% Graphite, 85% Vespel
Diameter (Metric):
For 0.1 to 0.25 mm ID column
For 0.1 to 0.32 mm ID column
For 0.32 mm ID column
For 0.45 to 0.53 mm ID column
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Diameter (Metric) Unit Size
10394473 15% Graphite, 85% Vespel For 0.45 to 0.53 mm ID column Pack of 10
10756334 100% Graphite For 0.1 to 0.32 mm ID column Pack of 10
10672674 100% Graphite For 0.45 to 0.53 mm ID column Pack of 10
10120513 15% Graphite, 85% Vespel For 0.1 to 0.25 mm ID column Pack of 10
10774827 15% Graphite, 85% Vespel For 0.32 mm ID column Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10394473 Supplier Thermo Scientific™ Supplier No. 290VA193
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 28-09-2026
Add to basket
Edge
Add to basket
Ensure reliable connections for Thermo Scientific™ TRACE™ 1300/1600 series and Agilent GC instruments with these Thermo Scientific™ Ferrules
Use Thermo Scientific™ Ferrules Instruments to ensure optimal performance of your Thermo Scientific and Agilent GC instrument. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications.
  • Available in 100% graphite ferrules, 15% graphite/85% vespel
  • Ferrules for different types of injectors and detectors
Ordering Information
Packaged in contaminant-free, individual blister packs.
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Diameter (Metric) For 0.45 to 0.53 mm ID column
For Use With (Equipment) SSL Injectors and detectors of Thermo Scientific TRACE 1300/1600 series or Agilent GC instruments
Material 15% Graphite, 85% Vespel
Quantity Pack of 10