Learn More
Ferroin Indicator Solution, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
- A high quality indicator solution such as Reagecon's Ferroin Indicator Solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Specifications
| Packaging | Amber glass Winchester bottle |
| Physical Form | Liquid |
| Technique | Titration, Analyte Color Change |
| Quantity | 100 mL |