missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferredoxin Reductase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 559.00€
Specifications
| Antigen | Ferredoxin Reductase |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625235
|
Novus Biologicals
NBP2-38530-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108828
|
Novus Biologicals
NBP2-38530 |
0.1 mL |
559.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferredoxin Reductase Polyclonal specifically detects Ferredoxin Reductase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Ferredoxin Reductase | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P22570 | |
| 2232 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MASRCWRWWGWSAWPRTRLPPAGSTPSFCHHFSTQEKTPQICVVGSGPAGFYTAQHLLKHPQAHVDIYEKQPVPFGLVRFGVAPDHPEVKNVINTFTQTAHSGRCAFWGNVEVGRDVTVPELREAYHAVVLSYG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Adrenodoxin reductase, ADXREC 1.18.1.2, ferredoxin reductaseAR, Ferredoxin--NADP(+) reductase, NADPH:adrenodoxin oxidoreductase, mitochondrial | |
| FDXR | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title