Learn More
Thermo Scientific™ Maxima™ Hot Start Green 2X PCR Master Mix
Product Code. p-4526941 Shop All Thermo Scientific Products
Click to view available options
Sufficient For:
100 reactions of 50 µl
500 reactions of 50 µl
Unit Size:
100 reactions
500 reactions
Product Code. 11893923
Brand: Thermo Scientific™ K1061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Save time preparing samples with this ready-to-use 2x hot start master mix containing tracking dyes and a density reagent for direct loading on gels after PCR.
Maxima Hot Start Green PCR Master Mix (2X) is a ready-to-use solution containing Maxima Hot Start Taq DNA Polymerase, optimized hot start PCR buffer, Mg2+, and dNTPs. The master mix is supplemented with two tracking dyes and a density reagent that allows for direct loading of PCR products on a gel.
The dyes in the master mix do not interfere with PCR performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion. The master mix retains all features of Maxima Hot Start Taq DNA Polymerase. It is capable of amplification of up to 3 kb from genomic DNA.
The master mix is supplemented with two tracking dyes and a density reagent that allows for direct loading of PCR products on a gel. The dyes in the master mix do not interfere with PCR performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion. The master mix retains all features of Maxima Hot Start Taq DNA Polymerase. It is capable of amplification of up to 3kb from genomic DNA. For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless Maxima Hot Start PCR Master Mix (2X) (†K0151).
  • Convenient—Maxima Hot Start Taq DNA Polymerase in a ready-to-use mix
  • Hot start PCR—reaction set-up at room temperature
  • Four minute activation time
  • Direct loading of PCR product on gels
  • Highs specificity and sensitivity of PCR
Recommended for:
High throughput hot start PCR; RT-PCR; Highly specific amplification of complex genomic and cDNA templates; Amplification of low copy DNA targets; Generation of PCR products for TA cloning
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fermentas™ Maxima™ Hot Start
Quantity 2 x 1.25mL
Sufficient For 100 reactions of 50 µl
For Use With (Application) Standard PCR, RT-PCR
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.