Learn More
Cytiva FEP Inlet Tubing Kit
Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
4.21 mm
Unit Size:
1 set
Specifications
Specifications
| Type | Inlet Tubing Kit |
| Includes | 1.5 m L 4.2 I.D. x 3.1 mm O.D. tubing, UNF 5/16 in. male 2 x nut, 2 x ferrule |
| Diameter (Metric) Inner | 4.21 mm |
| Material | FEP |