Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Fenpropathrin |
| CAS | 39515-41-8 |
| CAS Max % | ≤ 100% |
| Flash Point | 205°C |
| Linear Formula | C22H23NO3 |
| Beilstein | 2673776 |
| Color | Brown/Yellow |
| Melting Point | 45°C to 50°C |
| Synonym | cyano(3-phenoxyphenyl)methyl 2,2,3,3-tetramethylcyclopropanecarboxylate |
| Molecular Weight (g/mol) | 349.42 g/mol |
| Show More |