Learn More
Fenpropathrin, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (±)-Fenpropathrin |
| CAS | 39515-41-8 |
| Analyte or Component Names | (±)-Fenpropathrin |
| Shipping Condition | +20°C |
| Molecular Formula | C22 H23 N O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2,2,3,3-Tetramethylcyclopropanecarboxylic acid cyano(3-phenoxyphenyl)methyl ester, (±)-Fenpropathrin, 2,2,3,3-Tetramethylcyclopropanecarboxylic acid cyano(3-phenoxyphenyl)methyl ester, Danimen, Danitol, Danitol 10EC, Danitol 2.4EC, Danitol Fiori, Fenpropanate, Fenpropathrin, Kilumal, Meiothrin, Meothrin, Miothrin, Rody, S 3206, SD 41706, Smash, Tame, WL 41706, XE 938, α-Cyano-3-phenoxybenzyl 2,2,3,3-tetramethylcyclopropanecarboxylate |
| InChI Formula | InChI=1S/C22H23NO3/c1-21(2)19(22(21,3)4)20(24)26-18(14-23)15-9-8-12-17(13-15)25-16-10-6-5-7-11-16/h5-13,18-19H,1-4H3 |
| SMILES | CC1(C)C(C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2)C1(C)C |
| IUPAC Name | [cyano-(3-phenoxyphenyl)methyl] 2,2,3,3-tetramethylcyclopropane-1-carboxylate |
| Show More |