Learn More
Fenobucarb, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Fenobucarb |
| CAS | 3766-81-2 |
| Analyte or Component Names | Fenobucarb |
| Shipping Condition | +20°C |
| Molecular Formula | C12 H17 N O2 |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C12H17NO2/c1-4-9(2)10-7-5-6-8-11(10)15-12(14)13-3/h5-9H,4H2,1-3H3,(H,13,14) |
| SMILES | O=C(OC=1C=CC=CC1C(C)CC)NC |
| IUPAC Name | (2-butan-2-ylphenyl) N-methylcarbamate |
| Molecular Weight (g/mol) | 207.27 |
| Show More |