Learn More
Eppendorf™ Felt Tip Pen
Product Code. 12862849 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12862849 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12862849 Supplier Eppendorf™ Supplier No. 0013088101
Estimated Shipment: 16-11-2026
Mark tubes and other laboratory items with this pen. Eppendorf™ Felt Tip Pens are ideal for writing on a variety of surfaces.
  • Black Pen
  • For writing
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Application) Writing
Quantity 1 Ea.