missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FE65 Antibody (SR1014), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | FE65 |
|---|---|
| Clone | SR1014 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640784
|
Novus Biologicals
NBP3-22195-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631615
|
Novus Biologicals
NBP3-22195-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FE65 Monoclonal antibody specifically detects FE65 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| FE65 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein | |
| A synthesized peptide derived from human FE65 (Uniprot #: O00213) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1014 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 322 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title