missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRN/FCGRT Antibody (CL3640) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61627MFV610
This item is not returnable.
View return policy
Description
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FCRN/FCGRT | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| This antibody was developed using a recombinant protein derived from P55899, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Immunology | |
| 2217 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3640 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction