missing translation for 'onlineSavingsMsg'
Learn More

FCRN/FCGRT Antibody (CL3638) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30300781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300781 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30300781 Supplier Novus Biologicals Supplier No. NBP261626MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRN/FCGRT
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3638
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P55899, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2217
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.