missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FCRN/FCGRT Antibody (batoclimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28471-50ug
This item is not returnable.
View return policy
Description
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FCRN/FCGRT | |
| Monoclonal | |
| Unconjugated | |
| P55899 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| batoclimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| FcRn (FCGRT & B2M) | |
| 50 μg | |
| Immunology | |
| 2217 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction