missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCRL6 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149172
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Acts as a MHC class II receptor (PubMed:20519654). When stimulated on its own, does not play a role in cytokine production or the release of cytotoxic granules by NK cells and cytotoxic CD8(+) T cells (PubMed:17213291, PubMed:18991291). Does not act as an Fc receptor (PubMed:18991291). [UniProt].
Specifications
| FCRL6 | |
| Polyclonal | |
| Unconjugated | |
| FCRL6 | |
| Fc receptor homolog 6; Fc receptor like 6; Fc receptor-like 6; Fc receptor-like protein 6; Fc receptor-like protein 7; FCRH6; FCRL6; fcR-like protein 6; FLJ16056; IFGP6; IgSF type I transmembrane receptor; leukocyte receptor | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 343413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6DN72 | |
| FCRL6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6, Lys16-Leu312. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction