Learn More
FCRL4/FcRH4/IRTA1 Antibody (580810), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB24262C
Product Code. 18648933
  • 664.00€ / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
FCRL4/FcRH4/IRTA1 Monoclonal antibody specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FCRL4/FcRH4/IRTA1
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
83417
Store at 4°C in the dark.
IgG1
Flow Cytometry, CyTOF
580810
Flow Cytometry, CyTOF-ready
CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522
Mouse myeloma cell line NS0-derived recombinant human FCRL4/FcRH4, Gln16-Asp385, Accession # NP_112572
0.1 mL