missing translation for 'onlineSavingsMsg'
Learn More

FCER1G Antibody, FITC, Novus Biologicals™

Product Code. 18398295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398295 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398295 Supplier Novus Biologicals Supplier No. NBP306225F

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FCER1G Polyclonal specifically detects FCER1G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCER1G
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide, Fc-epsilon RI-gamma, FceRI gamma, FCRG, high affinity immunoglobulin epsilon receptor subunit gamma, IgE Fc receptor subunit gamma, immunoglobulin E receptor, high affinity, gamma chain
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FCER1G fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2207
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.