missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIA/CD32a Antibody (3B6), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Fc gamma RIIA/CD32a |
|---|---|
| Clone | 3B6 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043349
|
Novus Biologicals
NBP3-26587-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043171
|
Novus Biologicals
NBP3-26587-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Western Blot, ELISASpecifications
| Fc gamma RIIA/CD32a | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a | |
| A synthesized peptide derived from Human Fc gamma RIIA/CD32a [UniProt P12318] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3B6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2212 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title