missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RII/CD32 Mouse anti-Human, Clone: FCGR2A/479, Novus Biologicals™

Mouse Monoclonal Antibody

508.00€

Specifications

Antigen Fc gamma RII/CD32
Clone FCGR2A/479
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Functional
Applications Flow Cytometry, Immunofluorescence, Functional Assay
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18682918
View Documents
Novus Biologicals
NBP3-07227-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.
Specifications

Specifications

Fc gamma RII/CD32
0.2 mg/mL
Flow Cytometry, Immunofluorescence, Functional Assay
Unconjugated
Mouse
Human
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Recombinant human Fc gamma RII/CD32 protein
Primary
Store at 4°C.
FCGR2A/479
Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Functional
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
2213
IgG2b κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.