missing translation for 'onlineSavingsMsg'
Learn More

Fc epsilon RI Antibody (43160), HRP, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

593.00€

Specifications

Antigen Fc epsilon RI
Clone 43160
Applications Flow Cytometry
Classification Monoclonal
Conjugate HRP
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18578572
View Documents
Novus Biologicals
NBP1-43279H
0.1 mL
593.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications

Specifications

Fc epsilon RI
Flow Cytometry
HRP
Armenian Hamster
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
PBS
2205
IgG
Protein A or G purified
43160
Monoclonal
Purified
RUO
Mouse
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
The immunogen for this antibody was FceRI alpha.
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.