missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc epsilon RI alpha Antibody (Mar-1) - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
250.00€ - 680.00€
Specifications
| Antigen | Fc epsilon RI alpha |
|---|---|
| Clone | 2024-03-01 00:00:00 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635618
|
Novus Biologicals
NBP3-09039-0.025mg |
0.025 mg |
250.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671348
|
Novus Biologicals
NBP3-09039-0.2mg |
0.2 mg |
680.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc epsilon RI alpha Monoclonal antibody specifically detects Fc epsilon RI alpha in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
| Fc epsilon RI alpha | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| FceR1 alpha | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2024-03-01 00:00:00 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Armenian Hamster | |
| Mouse | |
| FCE1A;fcepsilonri;FCER1A;FcERI;Fcr-5 | |
| IgG λ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title