missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO22 Antibody (OTI1H1), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | FBXO22 |
|---|---|
| Clone | OTI1H1 |
| Dilution | Western Blot 1:500-2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306668
|
Novus Biologicals
NBP3-45022-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306339
|
Novus Biologicals
NBP3-45022-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXO22 Monoclonal antibody specifically detects FBXO22 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| FBXO22 | |
| Western Blot 1:500-2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| F-box only protein 22, F-box protein 22, F-box protein FBX22p44, FBX22FLJ13986, FIST domain containing 1, FISTC1, MGC31799 | |
| Human recombinant protein fragment corresponding to amino acids 1-220 of human FBXO22 (NP_671717) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI1H1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 26263 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title