missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38564-20ul
This item is not returnable.
View return policy
Description
FBXO2 Polyclonal antibody specifically detects FBXO2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| FBXO2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FBG1, F-box gene 1, F-box only protein 2, F-box protein 2, Fbs1, FBX2Fbg1, Nfb42, OCP1, organ of Corti protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human FBXO2 (NP_036300.2).,, Sequence:, MDGDGDPESVGQPEEASPEEQPEEASAEEERPEDQQEEEAAAAAAYLDELPEPLLLRVLAALPAAELVQACRLVCLRWKELVDGAPLWLLKCQQEGLVPEGGVEEERDHWQQFYFLSKRRRNLLRNPCGEEDLEGWCDVEHGGDGWRVEELPGDSGVEFTHDESVKKYFASSFEWCRKAQVIDLQAEGYWEELLDTTQPAIVVKDWYSGRSDAGCLYELTVKLLSEHENVLAEFSSGQVAVPQDSDGGGWMEISHTFTDYGPGVRFVRFEHGGQDSVYWKGWFGARVTNSSVWVEP | |
| 20 μL | |
| Cell Biology, Ubiquitin Proteasome Pathway | |
| 26232 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction