missing translation for 'onlineSavingsMsg'
Learn More

FBXL16 Antibody - BSA Free, Novus Biologicals™

Product Code. 18533243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18533243 0.1 mg 0.1mg
18562703 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18533243 Supplier Novus Biologicals Supplier No. NBP176527

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FBXL16 Polyclonal antibody specifically detects FBXL16 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL16
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS
Gene Accession No. NP_699181
Gene Alias C16orf22, c380A1.1, c380A1.1 (novel protein), chromosome 16 open reading frame 22, FBL16, F-box and leucine-rich repeat protein 16Fbl16, F-box/LRR-repeat protein 16, FLJ33735, MGC33974
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human FBXL16. The immunogen is located within the last 50 amino acids of FBXL16. Amino Acid Squence: PGATPELFKYFSQHLPR
Purification Method Peptide affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 146330
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.