missing translation for 'onlineSavingsMsg'
Learn More

Abcam Fast Green FCF, Tissue and Protein Stain, 25 g

Product Code. 32118453
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32118453 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32118453 Supplier Abcam Supplier No. ab14626725g
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Widely utilized tissue and protein stain for histones, electrophoresis, and Masson's trichrome applications.

  • Molecular Weight: 808.86 g/mol
  • Form: Solid
  • Applications: Tissue and protein stain, quantitative stain for histones at alkaline pH, protein stain in IEF and SDS-PAGE electrophoresis, food dye
  • Features: Fluoresces in near-infrared (NIR), replacement for Light Green SF yellowish in Masson's trichrome
  • Composition: 100% Fast green FCF

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Fast green FCF
Content And Storage Store at ambient temperature.
Detection Location In-Gel Detection
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Proteins, Histones
Detection Method Fluorescence
Formulation 100% Fast green FCF
Quantity 25 g
For Use With (Application) Tissue staining, protein staining, IEF, SDS-PAGE electrophoresis, Masson's trichrome staining, food coloring
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.