missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASN Mouse anti-Human, CoraLite 594, Clone: 1B6E4, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | FASN |
|---|---|
| Clone | 1B6E4 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17791419
|
Proteintech
CL594-66591-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17781419
|
Proteintech
CL594-66591-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FASN Monoclonal antibody specifically detects FASN in Human samples. It is validated for IFSpecifications
| FASN | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human | |
| 2194 | |
| Primary | |
| Store at -20°C |
| 1B6E4 | |
| Immunofluorescence | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG1 | |
| Protein G purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title