missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2, p Ser222 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
213.00€ - 450.00€
Specifications
| Antigen | FANCD2 (p Ser222) |
|---|---|
| Dilution | Western Blot 1:100-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18054365
|
Novus Biologicals
NB100-502SS |
0.05 mL |
213.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758263
|
Novus Biologicals
NB100-502 |
0.2 mL |
450.00€
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCD2 Polyclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot.Specifications
| FANCD2 (p Ser222) | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| Whole antisera with 0.1% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| FANCD2 | |
| IgG | |
| Unpurified | |
| 164.1 kDa |
| Western Blot 1:100-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q9BXW9 | |
| 2177 | |
| This FANCD2 [p Ser222] Antibody was developed against a synthetic peptide made to human FANCD2 (residues 217-226), surrounding the phosphorylated Serine 222. [Swiss-Prot# Q9BXW9] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title