missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
208.00€ - 322.00€
Specifications
| Antigen | FANCC |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634728
|
Novus Biologicals
NBP3-12777-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690228
|
Novus Biologicals
NBP3-12777-50ul |
50 μg |
208.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCC Polyclonal antibody specifically detects FANCC in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FANCC | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 0.2 um filtered solution in PBS | |
| 2176 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FA3, FACCProtein FACC, FACFanconi anemia group C protein, Fanconi anemia, complementation group C, FLJ14675 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FANCC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title