missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM59B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41118-0.025mg
This item is not returnable.
View return policy
Description
FAM59B Polyclonal specifically detects FAM59B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FAM59B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GAREM2 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FAM59B. The immunogen is located within amino acids 600 - 650 of FAM59B. Amino Acid Squence: PPLFKPSHPQKRFAPFGA | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| FAM59B, Family With Sequence Similarity 59, Member B, GAREML, GRB2 Associated, Regulator Of MAPK1-Like, GRB2-Associated And Regulator Of MAPK Protein-Like, GRB2-Associated And Regulator Of MAPK1-Like, KIAA2038, Protein FAM59B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 150946 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction