missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM43B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-05973CL1
This item is not returnable.
View return policy
Description
FAM43B Polyclonal antibody specifically detects FAM43B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| FAM43B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 163933 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| FAM43B family with sequence similarity 43, member B | |
| Produced in rabbits immunized with E. coli-derived Human FAM43B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction