missing translation for 'onlineSavingsMsg'
Learn More

FAM170A Antibody (OTI6E4), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44797

Product Code. 30306565

  • 735.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FAM170A Monoclonal antibody specifically detects FAM170A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM170A
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
340069
Human
Antibody
IgG1
Western Blot
OTI6E4
Western Blot 1:2000
family with sequence similarity 170, member A, Znf domain containing protein
Full length human recombinant protein of human FAM170A(NP_877438) produced in E.coli.
100 μg
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.