Description
- The 96-Well Dye Calibration Plate (FAM™) is used to maintain your real-time PCR instrument with standard 96-well block.
- Dye calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
Specifications
Specifications
| Format | 96-well Dye Calibration Plate |
| Content And Storage | Store at –20°C ± 5°C. |
| Calibrated Dye | FAM |
| For Use With (Equipment) | ViiA™ 7 Dx System, QuantStudio™ Dx |
| Reaction Speed | Standard |
| Product Type | Dye Calibration Plate |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 1 x 96-well plate |
| Calibration Type | Spectral Calibration |
| Show More |