missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor XI Recombinant Rabbit Monoclonal Antibody (007), HRP

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560261
This item is not returnable.
View return policy
Description
Factor XI Recombinant Monoclonal Antibody for ELISA
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality.
Specifications
| Factor XI | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% HRP Stabilizer and no preservative | |
| P03951 | |
| F11 | |
| Recombinant Human Coagulation Factor XI/F11 protein (Met1-Val625). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 007 | |
| HRP | |
| F11 | |
| 1600027G01Rik; AI503996; Cf11; Coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; FXI; LRRGT00086; plasma thromboplastin antecedent; PTA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2160 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction