missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FABP7/B-FABP Recombinant Protein Antigen

Product Code. 18660908 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1 mL
Product Code.
18660908 0.1 mL
1 options
This item is not returnable. View return policy

Product Code. 18660908

Brand: Novus Biologicals™ NBP252889PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 2173
Molecular Weight (g/mol) MolecularWeight-theroretical: 27 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
Gene Symbol FABP7
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-52889.It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking tide related information and a protocol, click here .
Research Category Cell Cycle and Replication, Stem Cells
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein FABP7/B-FABP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.