missing translation for 'onlineSavingsMsg'
Learn More

FABP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16817213 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16817213 20 μL 20µL
16807213 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16817213 Supplier Proteintech Supplier No. 106761AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fatty acid binding proteins (FABP) are small (approximately 13-14 kDa) intracellular proteins with a high degree of tissue specificity. FABPs are a class of cytoplasmic proteins that bind long chain fatty acids. They are abundantly present in various cell types and seem to play an important role in the intracellular utilization of fatty acids. There are at least six distinct types of FABP, each showing a specific pattern of tissue expression. FABP leaks, due to its small size, rapidly out of is chemically damaged dying cells leading to a rise in serum levels. Liver FABP is a sensitive marker for cell damage of liver cells in vitro and in vivo. Is chemically damaged tissues are characterized histologically by absence (or low presence) of FABP facilitating recognition of such areas. Next to this L FABP is a marker for rapid hepatocyte lysis in vitro (as for example in toxicology assays) and for detection of liver damage during and after transplantation.

Specifications

Antigen FABP3
Applications Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FABP3
Gene Accession No. P05413, P07483, P11404
Gene Alias FABP11, FABP3, Fatty acid binding protein 3, H FABP, HFABP, M FABP, MDGI, O FABP
Gene Symbols Fabp3
Host Species Rabbit
Immunogen FABP3 Fusion Protein Ag1069
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14077, 2170, 79131
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.