Learn More
IBA GmbH™ Fab-TACS™ Gravity Adapter
IBA GmbH™ Fab-TACS™ Gravity Adapter is designed to be used for reversible cell isolation delivering label-free target cells.
Brand: IBA GmbH™ 6-6331-001
Additional Details : Weight : 0.05000kg
Description
- Allows the fitting of Fab-TACS™ Gravity columns in standard 50mL tubes.
Specifications
| 1 adapter for 50mL tubes | |
| One adapter |