missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab)-Goat anti-Human IgG Fc Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 647

Product Code. 31369962
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31369962 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31369962 Supplier Invitrogen™ Supplier No. SA510372AFP647
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

F(ab)-Goat Recombinant Superclonal Secondary Antibody

The F(ab)-Goat anti-Human IgG Fc, Superclonal™ Recombinant Secondary Antibody is designed to specifically detect the Fc region of all Human IgG subtypes. It demonstrates moderate cross-reactivity with Rat and Monkey IgG, while showing no detectable reactivity with immunoglobulins from other tested species. This Fab fragment antibody is expressed in vitro and purified as a recombinant construct. It is also engineered to deliver enhanced stability and sensitivity. The advantages of recombinant antibodies include-better specificity, animal origin-free formulation, and lot-to-lot consistency. F(ab) fragments offer several advantages over full-length IgGs due to their smaller molecular size (~50 kDa vs. ~150 kDa), such as improved tissue penetration. The absence of the Fc region reduces nonspecific binding and background staining in Fc receptor-expressing cells and tissues, while also enabling increased flexibility in multiplexing assays. Alexa Fluor Plus dyes have been chemically optimized to provide higher signal to noise ratios and minimized cross-reactivity to help detect low-abundance targets in rare or precious samples. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG Fc
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG F(ab)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.