Learn More
EZH2/KMT6 Antibody (CL11945) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21205MFV610
Product Code. 30286807
  • 780.00€ / 0.10mL
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EZH2/KMT6
Monoclonal
mFluor Violet 610 SE
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
This antibody was developed using a synthetic peptide derived from Q15910, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer
2146
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11945
50mM Sodium Borate
Mouse
Protein A purified