Learn More
Invitrogen™ EXPRESS qPCR Supermix, universal
Click to view available options
No. of Reactions:
200 Reactions
2500 Reactions
Unit Size:
1 set
2500 reactions
Description
- EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays
- This kit comes with a separate tube of ROX reference dye
- EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available
This kit provides
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Specifications
| Content And Storage | EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Platinum Taq DNA Polymerase |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ |
| Passive Reference Dye | ROX (Separate Tube) |
| Product Type | Express qPCR Supermix |
| Show More |