missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOD1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97773PCP
This item is not returnable.
View return policy
Description
EXOD1 Polyclonal antibody specifically detects EXOD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| EXOD1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| EC 3.1, ERI1 exoribonuclease 2, ERI1 exoribonuclease family member 2, EXOD1, exonuclease domain containing 1, Exonuclease domain-containing protein 1, exoribonuclease 2, KIAA1504enhanced RNAi three prime mRNA exonuclease homolog 2, MGC16943 | |
| Produced in rabbits immunized with E. coli-derived Human EXOD1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 112479 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction