Learn More
Eppendorf™ Exchangeable Thermoblock
Product Code. p-8006963 Shop All Eppendorf Products
Change view
Click to view available options
Holds:
0.5 mL Tubes
1.5 mL tubes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10344651 0.5 mL Tubes Each
10439472 1.5 mL tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10344651 Supplier Eppendorf™ Supplier No. 5361000015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessories for ThermoStat plus/thermoMixer plus
  • Compact, bench top thermostat with thermoblocks for 0.2mL, 0.5mL, 1.0mL, 2.0mL tubes and microtitre plates designed to provide a high degree of flexibility for a multitude of applications
  • Programmable temperature and run time for special applications
  • GLP compliant
  • RS232 interface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 24 Reaction Vessels 0.5mL with Work Rack and Cool Pack 0°C
Holds 0.5 mL Tubes
No. of Places 24
Type Block