missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVX1 Monoclonal Antibody (OTI1A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811336S
This item is not returnable.
View return policy
Description
EVX1 Monoclonal antibody specifically detects EVX1 in Human samples. It is validated for Western BlotSpecifications
| EVX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P0DMM9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6818 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A9 | |
| Unconjugated | |
| SULT1A3 | |
| eve, even-skipped homeo box homolog 1; eve, even-skipped homeobox homolog 1; even skipped homeotic gene 1 homolog; even-skipped homeo box 1 (homolog of Drosophila); even-skipped homeobox 1; Evx1; EVX-1; Homeobox even-skipped homolog protein 1; NEWGENE_1565788 | |
| Full length human recombinant protein of human SULT1A3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction