Learn More
Eutech™ Refilling Reference Electrolyte (Double Junction pH electrode)
Product Code. 12973447
Change view
Click to view available options
Quantity:
480 mL
Unit Size:
480mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12973447 480 mL 480mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12973447 Supplier Thermo Scientific Eutech Supplier No. 01X211218
  • ONLINE EXCLUSIVE PRICE
    117.00€
    113.00€ / 480mL
    Save 4.00€
    3% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Refilling Reference Electrolyte
Description Saturated Potassium Chloride for Double Junction pH Electrodes
Solution Type Calibration Solution
Industry Type Laboratory Use
Quantity 480 mL