Learn More
Eutech™ pH Buffer Tablets
Description
Eutech™ pH buffer tablets provide a quick, convenient way to prepare pH buffers from tablets for pH calibrations. Each box contains 100 pH buffer tablets.
Features:
- 10 strips x 10 tablets per box
Specifications
Specifications
| Color | Green |
| For Use With (Application) | pH calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 10 |
| Physical Form | Tablet |
| Quantity | 100 Tablets |
| Solution Type | Buffer |